Skip to Content
Merck
All Photos(1)

Documents

330712X

Avanti

Ceramide LIPIDOMIX Mass Spec Standard

Avanti Research - A Croda Brand

Synonym(s):

LIPIDOMIX Mass Spec Standard, solution (1:1 dichloromethane:methanol)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

solution (1:1 dichloromethane:methanol)

packaging

ampule of 1 × 1 mL (330712X-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Packaging

2 mL amber glass sealed ampule

Components

The Ceramide LIPIDOMIX Mass Spec Standard is supplied in 1 mL of (1:1) dichloromethane:methanol and contains the following components:

Mixture ComponentTarget Conc. (μM)
  • C16 Ceramide (d18:1/16:0)30
  • C18 Ceramide (d18:1/18:0)15
  • C24 Ceramide (d18:1/24:0)75
  • C24:1 Ceramide (d18:1/24:1(15Z))37.5

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 860516PC16 Ceramide (d18:1/16:0), Avanti Research - A Croda Brand 16.1μg/mLSDS

  • 860518PC18 Ceramide (d18:1/18:0), Avanti Research - A Croda Brand 8.5μg/mLSDS

  • 860524PC24 Ceramide (d18:1/24:0), Avanti Research - A Croda Brand 48.8μg/mLSDS

  • 860525PC24:1 Ceramide (d18:1/24:1(15Z)), Avanti Research - A Croda Brand 24.3μg/mLSDS

also commonly purchased with this product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service