Skip to Content
Merck
All Photos(1)

Key Documents

C113040

Sigma-Aldrich

DMT-dC(ac) Phosphoramidite

configured for MerMade

Synonym(s):

DMT-dC(ac) Amidite, N-acetyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxycytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], N4-Acetyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]

Sign Into View Organizational & Contract Pricing

Select a Size

6 X 5 G
€1,470.00
6 X 10 G
€2,460.00

€1,470.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
6 X 5 G
€1,470.00
6 X 10 G
€2,460.00

About This Item

Empirical Formula (Hill Notation):
C41H50N5O8P
CAS Number:
Molecular Weight:
771.84
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

€1,470.00


Check Cart for Availability

Request a Bulk Order

biological source

non-animal source (no BSE/TSE risk)

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥98% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

packaging

pkg of 6 × 10 g C113040-6x10G (8 oz Bottle 28-400)

technique(s)

oligo synthesis: suitable

color

white to off-white

λ

conforms (UV/VIS Identity)

compatibility

configured for MerMade

nucleoside profile

base: deoxycytidine
base protecting group: acetyl
2' protecting group: none
5' protecting group: DMT
deprotection: fast/standard

storage temp.

-10 to -25°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=CC(NC(C)=O)=NC3=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C41H50N5O8P/c1-28(2)46(29(3)4)55(52-25-11-23-42)54-36-26-39(45-24-22-38(43-30(5)47)44-40(45)48)53-37(36)27-51-41(31-12-9-8-10-13-31,32-14-18-34(49-6)19-15-32)33-16-20-35(50-7)21-17-33/h8-10,12-22,24,28-29,36-37,39H,11,25-27H2,1-7H3,(H,43,44,47,48)/t36-,37+,39+,55?/m0/s1

InChI key

MECWEBCHRKVTNV-RLWDVSNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Proligo′s DNA phosphoramidites lead to high-yield and high-quality oligonucleotides due to their high coupling efficiency. The deprotection step of automated oligonucleotide synthesis is integral to synthesis time and final product quality. DMT-dC(ac) Phosphoramidite is an acetyl-protected DNA phosphoramidite used in fast deprotection chemistry for the rapid and high-yield synthesis of high-purity oligonucleotides. Key Features of dC(ac):
  • Ultrafast deprotection: 10 minutes at 65°C with AMA reagent
  • Key component in oligonucleotide synthesis
  • Industry-standard for fast deprotection chemistry
  • Operating for AMA deprotection methods without changes
  • Consistent lot-to-lot high purity and performance
  • Manufactured under a certified ISO 9001 quality systemThe product belongs to MerMade synthesizers.

Packaging

C113040-6x5G packaged in a 8 oz bottle with a 28-400 cap
C113040-6x10G packaged in a 8 oz bottle with a 28-400 cap

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service