Skip to Content
Merck
All Photos(1)

Key Documents

T111000

Sigma-Aldrich

DMT-dT Phosphoramidite

Synonym(s):

5′-O-(4,4′-dimethoxytrityl)-2′-deoxythymidine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite], 5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxythymidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], DMT-dT amidite, Thymidine, 5′-O-[bis(4-methoxyphenyl)phenylmethyl]-, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing

Select a Size

6 X 5 G
€297.00
6 X 10 G
€1,010.00
100 G
€1,540.00
4 X 30 G
€1,840.00
6 X 20 G
€1,960.00
186.2 G
€2,840.00
500 G
€6,760.00
20 X 40 G
€13,510.00
1 KG
€13,930.00

€297.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
6 X 5 G
€297.00
6 X 10 G
€1,010.00
100 G
€1,540.00
4 X 30 G
€1,840.00
6 X 20 G
€1,960.00
186.2 G
€2,840.00
500 G
€6,760.00
20 X 40 G
€13,510.00
1 KG
€13,930.00

About This Item

Empirical Formula (Hill Notation):
C40H49N4O8P
CAS Number:
Molecular Weight:
744.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.51

€297.00


Check Cart for Availability

Request a Bulk Order

biological source

non-animal source

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder or granules

technique(s)

oligo synthesis: suitable

impurities

≤0.3 wt. % water content (Karl Fischer)

color

white to off-white

λ

conforms (UV/VIS Identity)

nucleoside profile

base: deoxythymidine
base protecting group: none
2' protecting group: none
5' protecting group: DMT
deprotection: fast/standard

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=C(C)C(=O)NC3=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C40H49N4O8P/c1-27(2)44(28(3)4)53(50-23-11-22-41)52-35-24-37(43-25-29(5)38(45)42-39(43)46)51-36(35)26-49-40(30-12-9-8-10-13-30,31-14-18-33(47-6)19-15-31)32-16-20-34(48-7)21-17-32/h8-10,12-21,25,27-28,35-37H,11,23-24,26H2,1-7H3,(H,42,45,46)/t35-,36+,37+,53?/m0/s1

InChI key

UNOTXUFIWPRZJX-CEXSRUIHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-dT Phosphoramidite belongs to the DNA Phosphoramidites group. Some key features of standard protected DNA phosphoramidite are mentioned below:
  • Exocyclic amine functions are protected by a benzoyl group (dA(bz) anddC(bz)) or isobutyryl group (dG(ib))
  • Recommended cleavage and deprotection conditions are 8 hours at 55 °Cor 24 hours at room temperature using concentrated ammonia solution, for standard base-protected oligonucleotides
  • The high coupling efficiency of Proligo′s DNA phosphoramidites leads to high-yield and high-quality oligonucleotides

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service