Skip to Content
Merck
All Photos(1)

Key Documents

C0889

Sigma-Aldrich

Cytochalasin H from Phomopsis sp.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H39NO5
CAS Number:
Molecular Weight:
493.63
Beilstein:
1632647
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.77

form

crystals

storage temp.

2-8°C

SMILES string

C[C@H]1C\C=C\C2[C@H](O)C(=C)[C@@H](C)C3[C@H](Cc4ccccc4)NC(=O)[C@@]23C(OC(C)=O)\C=C\[C@](C)(O)C1

InChI

1S/C30H39NO5/c1-18-10-9-13-23-27(33)20(3)19(2)26-24(16-22-11-7-6-8-12-22)31-28(34)30(23,26)25(36-21(4)32)14-15-29(5,35)17-18/h6-9,11-15,18-19,23-27,33,35H,3,10,16-17H2,1-2,4-5H3,(H,31,34)/b13-9+,15-14+/t18-,19+,23?,24-,25+,26?,27+,29-,30+/m0/s1

InChI key

NAEWXXDGBKTIMN-QHDZSFFESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cytochalasin reported to have CNS activity

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ghaskadbi et al.
Experimental cell biology, 50(3), 155-161 (1982-01-01)
Cytochalasin H (CH) has been shown to inhibit the reaggregation of dissociated cells of a fresh water sponge. The effect is dose-dependent and reversible. Even after 24 h of CH treatment, the cells show the formation of pseudopodia suggesting that
Scott M Stoeger et al.
Cancer chemotherapy and pharmacology, 63(5), 807-818 (2008-07-29)
Previous studies have indicated that the ERK1/2 MAP kinase signaling pathway plays an important role not only in cell growth, cell cycle regulation, and differentiation, but also in determining the sensitivity of cells to anticancer agents as well. Furthermore, expression
S Ghaskadbi et al.
Toxicology, 33(3-4), 323-330 (1984-12-01)
Effects of the mould metabolite, cytochalasin H (CH) were studied in chick embryos explanted at stages 4, 5 and 8 and cultured in vitro. Morphological and histological observations revealed that the major effects of CH are inhibition of primary morphogenesis
P Natarajan et al.
Platelets, 11(8), 467-476 (2001-02-15)
Platelets contain a well-developed and dynamic cytoskeleton composed mainly of actin and actin-associated proteins. Upon platelet activation there is rapid polymerisation of actin and a marked reorganisation of the platelet cytoskeleton. Cytochalasins are agents that interfere with the polymerisation of
Effect of cytochalasins B & H on isoelectric focusing of chick embryonic neural retina cells.
S Ghaskadbi et al.
Indian journal of experimental biology, 20(12), 869-871 (1982-12-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service