Skip to Content
Merck
All Photos(1)

Key Documents

V5888

Sigma-Aldrich

VUF 5574

Synonym(s):

N-(2-Methoxyphenyl)-N′-[2-(3-pyridinyl)-4-quinazolinyl]-urea

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€196.00

€196.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€196.00

About This Item

Empirical Formula (Hill Notation):
C21H17N5O2
CAS Number:
Molecular Weight:
371.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€196.00


Estimated to ship on27 May 2025


Request a Bulk Order

form

solid

color

white

solubility

DMSO: soluble 1.5 mg/mL
H2O: insoluble

SMILES string

COc1ccccc1NC(=O)Nc2nc(nc3ccccc23)-c4cccnc4

InChI

1S/C21H17N5O2/c1-28-18-11-5-4-10-17(18)24-21(27)26-20-15-8-2-3-9-16(15)23-19(25-20)14-7-6-12-22-13-14/h2-13H,1H3,(H2,23,24,25,26,27)

InChI key

YRAFEJSZTVWUMD-UHFFFAOYSA-N

Gene Information

human ... ADORA3(140)

Biochem/physiol Actions

Potent, selective, and competitive A3 adenosine receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service