Skip to Content
Merck

Skip To

C0793

Fluorimeter cuvettes

Polymethacrylate, without stopper

Fluorimeter cuvettes

Synonym(s):

cuvette, plastic cuvette, plastic cuvettes

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 ea
Check Cart for Availability
€ 55,70

About This Item

NACRES:
SB.32
UNSPSC Code:
41121813

€ 55,70


Check Cart for Availability

Request a Custom OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

photometry: suitable

L × W × H

0.5 in. × 0.5 in. × 1.75 in.

volume

≥3 mL

detection method

fluorometric

General description

See transmission curve labelled C
These cuvettes have four optically clear sides.

Note: polystyrene and polymethacrylate cuvettes are incompatible with some organic solvents.

Other Notes

Not compatible with some organic solvents or strong acids.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C0918C5291C5416
volume

≥3 mL

volume

≥3 mL

volume

-

volume

-

L × W × H

0.5 in. × 0.5 in. × 1.75 in.

L × W × H

12.7 mm × 12.7 mm × 44.7 mm

L × W × H

-

L × W × H

-

detection method

fluorometric

detection method

fluorometric

detection method

photometric

detection method

photometric

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

UV/Vis spectroscopy: suitable, photometry: suitable

technique(s)

UV/Vis spectroscopy: suitable, photometry: suitable


Still not finding the right product?

Explore all of our products under Fluorimeter cuvettes




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the chemical compatibility? Not compatible with "some" organic solvents is not helpful. What about alcohols? Methanol? Ethanol? Isopropanol?

    1 answer
    1. These polymethylmethacrylate (PMMA) cuvettes are intended for use with aqueous solutions. The solutions can contain weak acids such as acetic acid or weak bases such as sodium bicarbonate, or they can contain detergents. Glycerol and ethylene glycol (or aqueous solutions of those compounds) can also be used. But the cuvettes are NOT compatible with chlorinated solvents such as chloroform or methylene chloride, esters such as ethyl acetate, ketones such as acetone, aromatics such as benzene or toluene, or low molecular weight alcohols such as methanol, ethanol or isopropanol. Information on the compatibility of PMMA with alkanes such as hexane, heptane or cyclohexane is inconsistent, so it would be best not to use those.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review