Skip to Content
Merck
All Photos(1)

Key Documents

BMSIPD960010BS

Sigma-Aldrich

Benchmark Stainless Steel Bulk Beads

bead diam. 10 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NA.82
Pricing and availability is not currently available.

material

stainless steel

packaging

pack of 500 g

manufacturer/tradename

Benchmark Scientific IPD9600-10BS

bead diam.

10 mm

compatibility

25mL stainless steel grinding jars

Looking for similar products? Visit Product Comparison Guide

General description

Pack of loose 10mm stainless steel grinding balls for use with tube homogenizers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service