This product is intended for use with the BenchMasher™ 400. Samples up to 400ml are loaded into the blender bags and are sealed upon closing the door. This is not a heat seal. Upon opening the door, the bag is open and the sample may be accessed. The bag is nonporous.
BMSIPD4400B400
Blender Bag
capacity 400 mL
Synonym(s):
homogenization bag, homogenizer bag
About This Item
Recommended Products
material
clear
low-density polyethylene
sterility
non-sterile
manufacturer/tradename
Benchmark Scientific IPD4400-B400
bag H × W
300 cm × 180 cm
capacity
400 mL
General description
Application
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How porous is this bag? What is its locking system?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service