Skip to Content
Merck
All Photos(1)

Key Documents

BAF134870100

Bel-Art® Write-on Label Tape

roll size 40 yard, blue, width 1 in., pk of 3

Synonym(s):

labelling tape, moisture resistant label tape, solvent resistant label tape

Sign Into View Organizational & Contract Pricing

Select a Size

3 EA
€72.20

€72.20


Check Cart for Availability


Select a Size

Change View
3 EA
€72.20

About This Item

UNSPSC Code:
55121611
NACRES:
NB.35

€72.20


Check Cart for Availability

material

blue

packaging

pk of 3

manufacturer/tradename

Bel-Art F13487-0100

roll size

40 yard

width

1 in.
25.4 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Select from six tape colors that correspond with familiar hazard color codes to highlight safety information written on the labels. Use yellow for chemical reactivity, green for safety labels, blue indicates health hazards, red alerts to flammability; orange indicates a biohazard and white can describe a specific hazard. The tape is custom manufactured from proprietary materials that resist moisture and most solvents. Will adhere to PTFE fluoropolymer resin covered materials and any clean surface. A maximum temperature of 247 degrees C for 30 minutes can be endured.

Features and Benefits

  • No sticky residue lingers when tape is stripped off
  • Marks made with pencil, ball-point pen or solvent ink marker survive most laboratory conditions

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Write-on is a trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service