Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
12 ea | Check Cart for Availability | € 22,30 € 20,07 |
About This Item
€ 20,07
List Price€ 22,30Save 10%material
high-density polyethylene bottle, polypropylene cap, polypropylene screw closure, square translucent bottle
joint
Joints threaded neck (28)
description
Not autoclavable
feature
with cap
packaging
pack of 12 ea
manufacturer/tradename
Nalgene® 2114-0002
bottle L × H × W
36 mm × 83 mm × 36 mm
capacity
60 mL
General description
Features and Benefits
- Wide mouth is easy to fill.
- Bottle design saves space in freezer.
- HDPE exhibits excellent chemical resistance to most acids, bases and alcohols, making these bottles suitable for a wide variety of uses.
- Linerless caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents.
- Leakproof† when used with Nalgene closures.
- Bottle and Carton are readily recyclable in most communities.
- Manufactured from non-toxic materials.
- Participates in My Green Lab′s ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| material high-density polyethylene bottle, polypropylene screw closure, polypropylene cap, square translucent bottle | material high-density polyethylene bottle, polypropylene cap, polypropylene screw closure, square translucent bottle | material high-density polyethylene bottle, polypropylene cap, polypropylene screw closure, square translucent bottle | material high-density polyethylene bottle, polypropylene cap, polypropylene screw closure, square translucent bottle |
| manufacturer/tradename Nalgene® 2114-0002 | manufacturer/tradename Nalgene® 2114-0006 | manufacturer/tradename Nalgene® 2114-0032 | manufacturer/tradename Nalgene® 2114-0008 |
| feature with cap | feature with cap | feature with cap | feature with cap |
| capacity 60 mL | capacity 175 mL | capacity 1,000 mL | capacity 250 mL |
| joint Joints threaded neck (28) | joint Joints threaded neck (38-415) | joint Joints threaded neck (63) | joint Joints threaded neck (43) |
| packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 6 ea | packaging pack of 12 ea |
Still not finding the right product?
Explore all of our products under Nalgene® bottles, style 2114
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
