Skip to Content
Merck
All Photos(1)

Key Documents

B6410

Sigma-Aldrich

Nalgene® bottles, style 2114

capacity 175 mL

Synonym(s):

HDPE bottles, nalgene bottle, nalgene bottles

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
€38.50

€38.50


Check Cart for Availability


Select a Size

Change View
12 EA
€38.50

About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

€38.50


Check Cart for Availability

material

high-density polyethylene bottle
polypropylene cap
polypropylene screw closure
square translucent bottle

Joint

Joints threaded neck (38-415)

description

Not autoclavable

feature

with cap

packaging

pack of 12 ea

manufacturer/tradename

Nalgene® 2114-0006

bottle diam. × H × W

51 mm × 106 mm × 51 mm

capacity

175 mL

Looking for similar products? Visit Product Comparison Guide

General description

Square, wide-mouth bottles

Features and Benefits

  • Exhibits chemical resistance to a wide range of acids, bases, and alcohols
  • Excellent for freezer storage up to −100°C (−148°F)
  • Easy to fill as it has a wide mouth
  • Designed to save space in the freezer
  • Made up of non-toxic materials
  • Nalgene closures enable leak-proof bottles that prevent contamination of reagents

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service