Skip to Content
Merck
All Photos(1)

Key Documents

B1042

Sigma-Aldrich

Benzyl alcohol

ACS reagent, ≥99%

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

ACS reagent

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

Assay

≥99%

autoignition temp.

817 °F

impurities

≤0.01% Benzaldehyde (C6H5CHO)
≤0.02% Acetophenone (C6H5COCH3)

ign. residue

≤0.005%

color

APHA: ≤20

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Béatrice Hechler et al.
The Journal of pharmacology and experimental therapeutics, 314(1), 232-243 (2005-03-29)
Our aim was to determine whether the newly described P2X1 antagonist NF449 [4,4',4'',4'''-(carbonylbis(imino-5,1,3-benzenetriylbis(carbonylimino)))tetrakis-benzene-1,3-disulfonic acid octasodium salt] could selectively antagonize the platelet P2X1 receptor and how it affected platelet function. NF449 inhibited alpha,beta-methyleneadenosine 5'-triphosphate-induced shape change (IC50 = 83 +/- 13
Yuma Morimoto et al.
Inorganic chemistry, 51(18), 10025-10036 (2012-09-08)
The rate of oxidation of 2,5-dimethoxybenzyl alcohol (2,5-(MeO)(2)C(6)H(3)CH(2)OH) by [Fe(IV)(O)(N4Py)](2+) (N4Py = N,N-bis(2-pyridylmethyl)-N-bis(2-pyridyl)methylamine) was enhanced significantly in the presence of Sc(OTf)(3) (OTf(-) = trifluoromethanesulfonate) in acetonitrile (e.g., 120-fold acceleration in the presence of Sc(3+)). Such a remarkable enhancement of the
Meenakshisundaram Sankar et al.
ACS nano, 6(8), 6600-6613 (2012-07-10)
We report a convenient excess anion modification and post-reduction step to the impregnation method which permits the reproducible preparation of supported bimetallic AuPd nanoparticles having a tight particle size distribution comparable to that found for sol-immobilization materials but without the
Luis E Santos-Figueroa et al.
Chemical Society reviews, 42(8), 3489-3613 (2013-02-13)
This review focuses on examples reported in the years 2010-2011 dealing with the design of chromogenic and fluorogenic chemosensors or reagents for anions.
I H Blank et al.
The Journal of investigative dermatology, 85(6), 522-526 (1985-12-01)
Although it is known that benzene may be absorbed from inhaled air, the amount that may enter the system by percutaneous absorption is less well established. We have measured the penetration of benzene through human abdominal skin in vitro from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service