Skip to Content
Merck

1.10164

SAFC

Hydrochloric acid 6 mol/L

EMPROVE® EXPERT

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301

Agency

CIP (cleaning in place)

Quality Level

product line

EMPROVE® EXPERT

form

liquid

concentration

≤0.0001% Cl- (Free Chlroine)
≤0.0001% Pb (Heavy metals)
≤0.00015% As (Arsenic)
≤0.0005% sulfate
5.7-6.3 mmol/L HCl

evapn. residue

≤0.01

pH

<1 (20 °C in H2O)

density

1.1 g/cm3 at 20 °C

application(s)

life science and biopharma (Biopharmaceutical Manufacturing)

storage temp.

2-25°C

Application

Filtered with 0.2 μmManufactured from Hydrochloric acid fuming 37% EMPROVE® ESSENTIAL PhEur, BP, JP, NF and purified water acc. PhEur, JP.Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service