Skip to Content
Merck
All Photos(1)

Key Documents

B9308

Sigma-Aldrich

BP897

>98% (HPLC), solid

Synonym(s):

N-[4-(4-(2-Methoxyphenyl)piperazinyl)butyl]-2-naphthamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€126.00

€126.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€126.00

About This Item

Empirical Formula (Hill Notation):
C26H31N3O2
CAS Number:
Molecular Weight:
417.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€126.00


Estimated to ship on22 May 2025


Request a Bulk Order

Assay

>98% (HPLC)

form

solid

color

off-white

solubility

DMSO: soluble 10 mg/mL

SMILES string

COc1ccccc1N2CCN(CCCCNC(=O)c3ccc4ccccc4c3)CC2

InChI

1S/C26H31N3O2/c1-31-25-11-5-4-10-24(25)29-18-16-28(17-19-29)15-7-6-14-27-26(30)23-13-12-21-8-2-3-9-22(21)20-23/h2-5,8-13,20H,6-7,14-19H2,1H3,(H,27,30)

InChI key

MNHDKMDLOJSCGN-UHFFFAOYSA-N

Biochem/physiol Actions

Partially selective D3 dopamine receptor agonist.

Legal Information

Sold with permission of INSERM and BIOPROJECT.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service