AXYRRP200CRS
Pipette tips for Packard® MPII and Janus®
robotic, racked, capacity 200 μL, clear, pkg of 5x960tips/cs, sterile
Synonym(s):
pipet tips
About This Item
Corning
sterility
sterile
manufacturer/tradename
Axygen Inc RRP-200-C-R-S
Corning
packaging
pkg of 5x960tips/cs
capacity
200 μL
volume
200 μL
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service