Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pipette tips for Packard
®
MPII and Janus
®
Products
Product Category
pipette tips (5)
Color
Manufacturer
Volume (µL)
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
AXYRRP200CBKR
Pipette tips for Packard
®
MPII and Janus
®
,
conductive, racked, capacity 200 μL, black, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYRRP50CBKR
Pipette tips for Packard
®
MPII and Janus
®
,
conductive, racked, capacity 50 μL, black, pkg of 5x960tips/cs
Expand
View Pricing
AXYRRP20CR
Pipette tips for Packard
®
MPII and Janus
®
,
racked, capacity 20 μL, pkg of 5x960tips/cs
Expand
View Pricing
AXYRRP200CRS
Pipette tips for Packard
®
MPII and Janus
®
,
robotic, racked, capacity 200 μL, clear, pkg of 5x960tips/cs, sterile
Expand
View Pricing
AXYRRP50CR
Pipette tips for Packard
®
MPII and Janus
®
,
racked, capacity 50 μL, clear, pkg of 5x960tips/cs
Expand
View Pricing