Documents
AXYP5ML48C100
Corning® Axygen® 48 Rectangular Well Deep Well Plate
well volume (5 mL), pyramid bottom, case of 100 ea
Synonym(s):
48-Well Deep Well Plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
pyramid bottom
packaging
case of 100 ea
manufacturer/tradename
Corning P-5ML-48-C-100
well volume
(5 mL)
Related Categories
General description
  • Axygen deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
  • Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80 C.
  • <UL><LI>5.0 mL deep
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.