Skip to Content
Merck
All Photos(2)

Key Documents

AV53829

Sigma-Aldrich

Anti-PAOX (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Polyamine oxidase (exo-N4-amino)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

25 kDa

species reactivity

horse, dog, mouse, rat, rabbit, human, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAOX(196743)

Immunogen

Synthetic peptide directed towards the C terminal region of human PAOX

Application

Anti-PAOX (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

PAOX [polyamine oxidase (exo-N4-amino)] encodes a FAD containing enzyme that catalyzes the oxidation of N(1)-acetylspermine and N1-acetylspermidine to spermidine and putrescine, respectively. It is, therefore, involved in the polyamine back-conversion. It also facilitates the regulation of polyamine intracellular concentration.

Sequence

Synthetic peptide located within the following region: LCLTQVLRRVTGNPRLPAPKSVLRSRWHSAPYTRGSYSYVAVGSTGGDLD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Bjelakovic et al.
Molecular and cellular biochemistry, 341(1-2), 79-85 (2010-04-21)
Diabetes mellitus is a metabolic disease characterized by inadequate secretion of insulin. Polyamine oxidase (PAO), a FAD-containing enzyme is involved in the biodegradation of Sp and Spd, catalyzing the oxidative deamination of Sp and Spd, resulting in production of ammonia
Aki Järvinen et al.
The Journal of biological chemistry, 281(8), 4589-4595 (2005-12-16)
FAD-dependent polyamine oxidase (PAO; EC 1.5.3.11) is one of the key enzymes in the catabolism of polyamines spermidine and spermine. The natural substrates for the enzyme are N1-acetylspermidine, N1-acetylspermine, and N1,N12-diacetylspermine. Here we report that PAO, which normally metabolizes achiral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service