Skip to Content
Merck
All Photos(2)

Documents

AV53693

Sigma-Aldrich

Anti-HSPA4L antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-APG-1, Anti-Heat shock 70 kDa protein 4-like, Anti-Osp94

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

95 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPA4L(22824)

General description

Heat shock 70kDa protein 4L is a protein encoded by the HSPA4L gene in humans. Heat shock proteins HSPA4L and HSPA4 are closely related members of the HSP110 family and act as cochaperones.

Immunogen

Synthetic peptide directed towards the C terminal region of human HSPA4L

Application

Anti-HSPA4L antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/mL.

Biochem/physiol Actions

Hspa4l is expressed ubiquitously and predominantly in the testis and is highly expressed in spermatogenic cells. It plays an important role in osmotolerance. HSPA4L and HSPA4 collaborate in embryonic lung maturation and helps in air breathing during child birth. Osp94 also acts as molecular chaperone and possesses cytoprotective role from excessive stimulation from heat, hyper-ionic and osmotic stress which cause marked perturbation of intracellular protein function including the suppression of protein synthesis.

Sequence

Synthetic peptide located within the following region: KDERYDHLDPTEMEKVEKCISDAMSWLNSKMNAQNKLSLTQDPVVKVSEI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Yamamoto et al.
Neuroscience, 158(4), 1691-1698 (2008-12-09)
Osmotic stress protein 94 (OSP94), a member of the heat shock protein 110/SSE subfamily, is expressed in certain organs such as the kidney, testis, and brain where it can act as a molecular chaperon. In general, its alteration in expression
Belal A Mohamed et al.
American journal of respiratory cell and molecular biology, 50(4), 817-824 (2013-08-29)
Heat shock proteins HSPA4L and HSPA4 are closely related members of the HSP110 family and act as cochaperones. We generated Hspa4l(-/-)Hspa4(-/-) mice to investigate a functional complementarity between HSPA4L and HSPA4 during embryonic development. Hspa4l(-/-)Hspa4(-/-) embryos exhibited marked pulmonary hypoplasia
Torsten Held et al.
Molecular and cellular biology, 26(21), 8099-8108 (2006-08-23)
The Hspa4l gene, also known as Apg1 or Osp94, belongs to the HSP110 heat shock gene family, which includes three genes encoding highly conserved proteins. This study shows that Hspa4l is expressed ubiquitously and predominantly in the testis. The protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service