Skip to Content
Merck
All Photos(1)

Key Documents

AV49489

Sigma-Aldrich

Anti-FAM20C (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DMP4, Anti-Family with sequence similarity 20, member C, Anti-RNS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

66 kDa

species reactivity

rat, guinea pig, rabbit, dog, mouse, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAM20C(56975)

General description

FAM20C (Family with sequence similarity 20, member C) is a member of FAM20 protein family. It is distributed in several mammalian cell lines. During hematopoietic differentiation, it is expressed in hematopoietic cells.

Immunogen

Synthetic peptide directed towards the middle region of human FAM20C

Application

Anti-FAM20C (AB1) antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

FAM20C (Family with sequence similarity 20, member C) is a secretory Golgi casein kinase involved in biomineralization, enamel formation, lipid homeostasis, wound healing, cell migration and adhesion. It has the ability to phosphorylate S-x-E/pS motifs on proteins in milk and in the extracellular matrix of bones and teeth. During enamel formation, it forms a functional complex by binding to a pseudokinase, Fam20A, which triggers extracellular protein phosphorylation within the secretory pathway. Mutations in FAM20C gene cause Raine syndrome, hypophosphatemia, hyperphosphaturia, dental anomalies, intracerebral calcifications and osteosclerosis.

Sequence

Synthetic peptide located within the following region: CFYGECSYYCSTEHALCGKPDQIEGSLAAFLPDLSLAKRKTWRNPWRRSY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exome sequencing reveals FAM20c mutations associated with fibroblast growth factor 23-related hypophosphatemia, dental anomalies, and ectopic calcification.
Rafaelsen SH, et al.
Journal of Bone and Mineral Research, 28, 1378-1385 (2013)
Demet Nalbant et al.
BMC genomics, 6, 11-11 (2005-01-29)
Hematopoiesis is a complex developmental process controlled by a large number of factors that regulate stem cell renewal, lineage commitment and differentiation. Secreted proteins, including the hematopoietic growth factors, play critical roles in these processes and have important biological and
Vincent S Tagliabracci et al.
Cell, 161(7), 1619-1632 (2015-06-20)
The existence of extracellular phosphoproteins has been acknowledged for over a century. However, research in this area has been undeveloped largely because the kinases that phosphorylate secreted proteins have escaped identification. Fam20C is a kinase that phosphorylates S-x-E/pS motifs on
Hypophosphatemic osteomalacia and bone sclerosis caused by a novel homozygous mutation of the FAM20C gene in an elderly man with a mild variant of Raine syndrome.
Takeyari S, et al.
Bone, 67, 56-62 (2014)
Jixin Cui et al.
eLife, 4, e06120-e06120 (2015-03-20)
Although numerous extracellular phosphoproteins have been identified, the protein kinases within the secretory pathway have only recently been discovered, and their regulation is virtually unexplored. Fam20C is the physiological Golgi casein kinase, which phosphorylates many secreted proteins and is critical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service