Skip to Content
Merck
All Photos(2)

Key Documents

AV44701

Sigma-Aldrich

Anti-MGAT2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CDGS2, Anti-GLCNACTII, Anti-GNT-II, Anti-GNT2, Anti-Mannosyl (α-1,6-)-glycoprotein β-1,2-N-acetylglucosaminyltransferase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€385.00

€385.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€385.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€385.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

51 kDa

species reactivity

rat, mouse, human, rabbit, horse, dog, bovine, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MGAT2(4247)

Immunogen

Synthetic peptide directed towards the middle region of human MGAT2

Application

Anti-MGAT2 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

Mannosyl (α-1,6-)-glycoprotein β-1,2-N-acetylglucosaminyltransferase (MGAT2) is a Golgi enzyme highly expressed in small intestine of mice and humans. It is a key regulator of fat metabolism and energy expenditure. Mutations in this gene show resistance to metabolic disorders induced by high-fat feeding and exhibit increased energy expenditure.

Sequence

Synthetic peptide located within the following region: PKNAALKLGCINAEYPDSFGHYREAKFSQTKHHWWWKLHFVWERVKILRD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David W Nelson et al.
Journal of lipid research, 52(9), 1723-1732 (2011-07-08)
Acyl CoA:monoacylglycerol acyltransferase 2 (MGAT2) is thought to be crucial for dietary fat absorption. Indeed, mice lacking the enzyme (Mogat2(-/-)) are resistant to obesity and other metabolic disorders induced by high-fat feeding. However, these mice absorb normal quantities of fat.
Takuma Tsuchida et al.
Lipids in health and disease, 11, 75-75 (2012-06-16)
Resynthesis of triglycerides in enterocytes of the small intestine plays a critical role in the absorption of dietary fat. Acyl-CoA:monoacylglycerol acyltransferase-2 (MGAT2) is highly expressed in the small intestine and catalyzes the synthesis of diacylglycerol from monoacylglycerol and acyl-CoA. To
Chi-Liang Eric Yen et al.
Nature medicine, 15(4), 442-446 (2009-03-17)
Animals are remarkably efficient in absorbing dietary fat and assimilating this energy-dense nutrient into the white adipose tissue (WAT) for storage. Although this metabolic efficiency may confer an advantage in times of calorie deprivation, it contributes to obesity and associated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service