Skip to Content
Merck

Skip To

AV13104

Anti-TRHR antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Thyrotropin-releasing hormone receptor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
€ 340,00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
2

€ 340,00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

45 kDa

species reactivity

rat, rabbit, mouse, human, pig, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRHR(7201)

Immunogen

Synthetic peptide directed towards the middle region of human TRHR

Application

Anti-TRHR antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

Thyrotropin-releasing hormone/thyrotropin-releasing factor (TRH, TRF) is a tripeptidal hormone that binds to Thyrotropin-releasing hormone receptor. It subsequently stimulates the synthesis and release of TSH (thyroid-stimulating hormone) and prolactin from the anterior pituitary.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: ISCGYKISRNYYSPIYLMDFGVFYVVPMILATVLYGFIARILFLNPIPSD

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB3500567AV33885AV46022
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

IgG fraction of antiserum

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

species reactivity

rat, rabbit, mouse, human, pig, dog

species reactivity

mouse, human

species reactivity

dog, human, horse, rat, guinea pig, rabbit

species reactivity

bovine, mouse, horse, human, rabbit, dog, rat, guinea pig


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters