Skip to Content
Merck
All Photos(1)

Key Documents

A4266

Sigma-Aldrich

Acetaldehyde-d4

Synonym(s):

Tetradeuteroacetaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CDO
CAS Number:
Molecular Weight:
48.08
Beilstein:
1743039
MDL number:
UNSPSC Code:
12352103

vapor pressure

14.05 psi ( 20 °C)

autoignition temp.

347 °F

refractive index

n20/D 1.3321 (lit.)

bp

21 °C (lit.)

mp

-125 °C (lit.)

mass shift

M+4

storage temp.

−20°C

SMILES string

[2H]C(=O)C([2H])([2H])[2H]

InChI

1S/C2H4O/c1-2-3/h2H,1H3/i1D3,2D

InChI key

IKHGUXGNUITLKF-MZCSYVLQSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecilia Sala et al.
European journal of mass spectrometry (Chichester, England), 23(2), 64-69 (2017-06-29)
Recently, the formation of carbonyl compound within e-cigarettes usage has been reported. The aim of this study was to develop a new analytical method for the direct analysis of carbonyl compounds in vaporized liquids. Two different types of e-cigarettes and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service