Skip to Content
Merck

Skip To

A3017

Aluminum chloride hexahydrate

≥98%

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
AlCl3 · 6H2O
CAS Number:
Molecular Weight:
241.43
EC Number:
231-208-1
UNSPSC Code:
12352103
MDL number:


vapor pressure

1 mmHg ( 100 °C)

assay

≥98%

SMILES string

Cl[Al](Cl)Cl.O.O.O.O.O.O

InChI

1S/Al.3ClH.6H2O/h;3*1H;6*1H2/q+3;;;;;;;;;/p-3

InChI key

JGDITNMASUZKPW-UHFFFAOYSA-K



Still not finding the right product?

Explore all of our products under Aluminum chloride hexahydrate


pictograms

Health hazardCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

target_organs

Lungs

supp_hazards

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library