Skip to Content
Merck

1.01084

SAFC

Aluminium chloride hexahydrate

EMPROVE® ESSENTIAL, Ph. Eur., BP, USP

Pharma Manufacturing

Synonym(s):

Aluminium chloride hexahydrate, hydrochloric acid aluminium salt hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AlCl3 · 6H2O
CAS Number:
Molecular Weight:
241.43
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-208-1

Agency

BP
Ph. Eur.
USP

Quality Level

description

EMPROVE® ESSENTIAL Ph Eur,BP,USP

product line

EMPROVE® ESSENTIAL

form

solid

potency

3311 mg/kg LD50, oral (Rat)

pH

2.5 (20 °C, 50 g/L in H2O)

mp

100 °C

density

2.40 g/cm3 at 20 °C

application(s)

liquid formulation
pharmaceutical
semi-solid formulation

storage temp.

15-25°C

InChI

1S/Al.3ClH.6H2O/h;3*1H;6*1H2/q+3;;;;;;;;;/p-3

InChI key

JGDITNMASUZKPW-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Aluminium chloride is a commonly used ingredient in pharmaceutical applications. Within these applications, Aluminium chloride has been used primarily in topic formulations.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Target Organs

Lungs

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service