Skip to Content
Merck

Skip To

A1972

[Asn670, Leu671]-Amyloid β/A4 Precursor Protein 770 Fragment 667-676 trifluoroacetate salt

≥95% (HPLC), solid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 149,00

About This Item

Empirical Formula (Hill Notation):
C50H78N14O19
CAS Number:
Molecular Weight:
1179.24
UNSPSC Code:
12352209
NACRES:
NA.32
MDL number:
Assay:
≥95% (HPLC)
Form:
solid

€ 149,00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APP(351)

Biochem/physiol Actions

Substrate for assaying β-secretase activity that corresponds to the "Swedish" Lys-Met/Asn-Leu mutation of the amyloid precursor protein β-secretase cleavage site.
Substrate for assaying β-secretase activity; "Swedish" Lys-Met/Asn-Leu mutation of amyloid precursor protein β-secretase cleavage site.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A3597A2201565740
description

≥95% (HPLC), solid

description

≥95% (HPLC), solid

description

≥95% (HPLC)

description

A cell-permeable benzolactam derived PKC activator (Ki = 11.9 nM for PKCα) that efficiently enhances non-amyloidogenic α-processing of amyloid precursor protein (APP) even at 100 nM in human fibroblast AG06848.

form

solid

form

solid

form

powder

form

solid

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

UniProt accession no.

P05067

UniProt accession no.

P01189

UniProt accession no.

P05067

UniProt accession no.

-

Gene Information

human ... APP(351)

Gene Information

human ... POMC(5443)

Gene Information

human ... APP(351)

Gene Information

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters