Skip to Content
Merck
All Photos(1)

Key Documents

A3597

Sigma-Aldrich

[D-Lys16]-Adrenocorticotropic Hormone Fragment 1-24 human, rat trifluoroacetate salt

≥95% (HPLC), solid

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€307.00

€307.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
500 μG
€307.00

About This Item

Empirical Formula (Hill Notation):
C136H210N40O31S
Molecular Weight:
2933.44
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
biological source:
human
form:
solid

€307.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

human

Quality Level

Assay

≥95% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

SMILES string

FC(F)(F)C(=O)O.S(CC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)CO)Cc9ccc(cc9)O)CO)C(=O)N[C@@H](CCC(=O)O)C(=O)N[C@@H](Cc8nc[nH]c8)C(=O)N[C@@H](Cc7ccccc7)C(=O)N[C@@H](CCCNC(=N)N)C(=O)N[C@@H](Cc5c6c([nH]c5)cccc6)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N1[C@@H](CCC

InChI

1S/C136H210N40O31S.C2HF3O2/c1-75(2)109(127(200)154-71-106(181)156-88(31-13-17-52-137)114(187)158-89(32-14-18-53-138)115(188)159-91(35-21-56-149-134(142)143)116(189)164-96(37-23-58-151-136(146)147)131(204)175-60-25-39-104(175)126(199)173-111(77(5)6)128(201

InChI key

MCAQPIPWSBBXDA-MMNZLOBGSA-N

Gene Information

human ... POMC(5443)

Amino Acid Sequence

Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro

Biochem/physiol Actions

ACTH agonist; used in the study of cortisol production.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service