Skip to Content
Merck
All Photos(1)

Key Documents

90819

Sigma-Aldrich

Bis(2,2′-bipyridine)-(5-aminophenanthroline)ruthenium bis(hexafluorophosphate)

BioReagent, suitable for fluorescence

Synonym(s):

Ru(bpy)2(phen-5-NH2)(PF6)2

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€255.00

€255.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€255.00

About This Item

Empirical Formula (Hill Notation):
C32H25N7Ru · 2 PF6
CAS Number:
Molecular Weight:
898.59
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.21

€255.00


Check Cart for Availability

Request a Bulk Order

product line

BioReagent

form

powder

reaction suitability

core: ruthenium
reaction type: Photocatalysis
reagent type: catalyst

fluorescence

λex 458 nm; λem 612 nm in 0.1 M Tris pH 7.0

suitability

suitable for fluorescence

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

[Ru++].F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.c1ccc(nc1)-c2ccccn2.c3ccc(nc3)-c4ccccn4.Nc5cc6cccnc6c7ncccc57

InChI

1S/C12H9N3.2C10H8N2.2F6P.Ru/c13-10-7-8-3-1-5-14-11(8)12-9(10)4-2-6-15-12;2*1-3-7-11-9(5-1)10-6-2-4-8-12-10;2*1-7(2,3,4,5)6;/h1-7H,13H2;2*1-8H;;;/q;;;2*-1;+2

InChI key

QQMMCXKCYXQNNA-UHFFFAOYSA-N

Application

Used for photoinduced intramolecular electron transfer in peptide-bridged molecules.

Features and Benefits

Sigma-Aldrich offers a series of reactive dyes based on ruthenium complexes for use in fluorescence polarization assays and fluorescence lifetime imaging applications. They have been designed specifically to have long fluorescence lifetimes. These dyes were first synthesized in the labs of Dr. Joseph Lakowicz at the Center for Fluorescence Microscopy. These laser-excitable metal ligand complexes have fluorescence lifetimes near 500 nanoseconds.
These dyes can be used to label amines on biomolecules under mild conditions. Advantages of these dyes as protein labels include high photostability, good water solubility, a lack of dye-dye interactions and large Stokes′ shifts.
In addition, the fluorescence signal of long-lived fluorophores can be gated to eliminate the emission from short-lifetime fluorophores and autofluorescence from cells and biomolecules to further improve sensitivity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Photoinduced intramolecular electron transfer in peptide-bridged molecules[1][2]

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.S. Schanze et al.
Journal of the American Chemical Society, 110, 1180-1180 (1988)
E Terpetschnig et al.
Analytical biochemistry, 251(2), 241-245 (1997-09-23)
We report the synthesis and spectral properties of a long-lifetime luminescent Ru complex containing a sulfhydryl-reactive maleimide group, [Ru (2,2'-bipyridine)2(1, 10-phenanthroline-5-maleimide)](PF6)2. [Ru(bpy)2(phen-mi)]2+ was covalently linked to human serum albumin, immunoglobulin G, and beta-galactosidase. The lifetimes for probe bound to proteins
Tao Wang et al.
Analytica chimica acta, 1062, 124-130 (2019-04-06)
Vibrio parahaemolyticus (VP), as a kind of harmful bacterium to human beings, its rapid and accurate detection is particularly important. Herein, we developed a simple and efficient dual-modal Faraday cage-type immunosensor for ultrasensitive detection of VP based on electrochemiluminescence (ECL)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service