Skip to Content
Merck
All Photos(3)

Key Documents

904945

Sigma-Aldrich

[Ru(dmbpy)3](PF6)2

≥95%

Synonym(s):

Tris(4,4′-dimethyl-2,2′-bipyridine)ruthenium bis(hexafluorophosphate)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C36H36F12N6P2Ru
CAS Number:
Molecular Weight:
943.71
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Photocatalysis

mp

>300 °C

SMILES string

[Ru].n5c(cc(cc5)C)c6nccc(c6)C.n3c(cc(cc3)C)c4nccc(c4)C.n1c(cc(cc1)C)c2nccc(c2)C.C

InChI

1S/3C12H12N2.CH4.Ru/c3*1-9-3-5-13-11(7-9)12-8-10(2)4-6-14-12;;/h3*3-8H,1-2H3;1H4;

InChI key

YXZDNATXDNOJQJ-UHFFFAOYSA-N

Application

This ruthenium photocatalyst facilitates a variety of transformations using visible light.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yusuke Tamaki et al.
Chemical science, 6(12), 7213-7221 (2015-12-01)
We discovered an extremely suitable sacrificial electron donor, 1,3-dimethyl-2-(o-hydroxyphenyl)-2,3-dihydro-1H-benzo[d]imidazole, for the selective photocatalytic reduction of CO2 to formic acid using a Ru(ii)-Ru(ii) supramolecular photocatalyst. The efficiency, durability, and rate of photocatalysis are significantly increased (ΦHCOOH = 0.46, TONHCOOH = 2766
Zhaofeng Wang et al.
Nature, 554(7690), 86-91 (2018-02-02)
Carbon has the unique ability to bind four atoms and form stable tetravalent structures that are prevalent in nature. The lack of one or two valences leads to a set of species-carbocations, carbanions, radicals and carbenes-that is fundamental to our

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service