Skip to Content
Merck
All Photos(1)

Key Documents

56963

Sigma-Aldrich

Atto Thio12

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€454.00

€454.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€454.00

About This Item

MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

€454.00


Estimated to ship on20 May 2025


Request a Bulk Order

product line

BioReagent

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

ATTO-TEC GmbH

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 575-581 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

[O-]Cl(=O)(=O)=O.CN(C)c1ccc2c(SC3=C\C(C=CC3=C2c4ccccc4C(=O)N(C)CCCC(O)=O)=[N+](\C)C)c1

InChI

1S/C29H31N3O3S.ClHO4/c1-30(2)19-12-14-23-25(17-19)36-26-18-20(31(3)4)13-15-24(26)28(23)21-9-6-7-10-22(21)29(35)32(5)16-8-11-27(33)34;2-1(3,4)5/h6-7,9-10,12-15,17-18H,8,11,16H2,1-5H3;(H,2,3,4,5)

InChI key

CNHBWUJLSIHMSI-UHFFFAOYSA-N

Application

Atto Thio12 is a new label closely related to the well-known rhodamines for coupling to biomolecules such as DNA, RNA or proteins. The characteristic feature of the label is high yield of triplet formation. The dye is moderately hydrophilic.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcia Levitus et al.
Quarterly reviews of biophysics, 44(1), 123-151 (2010-11-27)
The breakthroughs in single molecule spectroscopy of the last decade and the recent advances in super resolution microscopy have boosted the popularity of cyanine dyes in biophysical research. These applications have motivated the investigation of the reactions and relaxation processes
Andriy Chmyrov et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 7(11), 1378-1385 (2008-10-30)
Photo-induced switching of dyes into dark, long-lived states, such as a triplet state, has recently gained increasing interest, as a means to achieve ultra-high optical resolution. Additionally, these long lived states are often highly environment-sensitive and their photodynamics can thus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service