Skip to Content
Merck
All Photos(1)

Key Documents

50492

Sigma-Aldrich

Atto Rho101 NHS ester

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€411.00

€411.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€411.00

About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

€411.00


Check Cart for Availability

Request a Bulk Order

product line

BioReagent

Assay

90% (Coupling efficiency with benzylamine, HPLC)

form

solid

manufacturer/tradename

ATTO-TEC GmbH

transmittance

254 nm
586 nm

fluorescence

λex 586 nm; λem 610 nm

λ

in ethanol (0.1% trifluoroacetic acid)

suitability

suitable for fluorescence

storage temp.

−20°C

Application

Atto Rho101 NHS ester is a derivative of the well-known dye Rhodamine 101 for coupling to bio-molecules such as DNA, RNA or proteins. Characteristic features of the label are strong absorption, extraordinarily high fluorescence quantum yield, and high thermal and photo-stability. The dye is moderately hydrophilic. ATTO Rho101 is a cationic dye. After coupling to a substrate the dye carries a net electrical charge of +1.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-lin Jiang et al.
Guang pu xue yu guang pu fen xi = Guang pu, 31(5), 1348-1351 (2011-08-02)
The absorption, fluorescence and time-resolved fluorescence spectra of Rhodamine 101 dye in both methanol and acidic methanol solutions were measured. The authors achieved the characteristic information of the absorption and fluorescence spectra, and obtained the S1 lifetimes. The authors assigned
Kakishi Uno et al.
Journal of the American Chemical Society, 133(34), 13558-13564 (2011-08-09)
Photoswitchable or photoactivatable fluorescent dyes are potentially applicable to ultrahigh density optical memory media as well as super-resolution fluorescence imaging when the dyes are highly fluorescent and have large absorption coefficients. Here, we report on highly fluorescent photochromic dyes, which
Knut Rurack et al.
Analytical chemistry, 83(4), 1232-1242 (2011-01-22)
The determination of the fluorescence quantum yields (QY, Φ(f)) of a series of fluorescent dyes that span the absorption/excitation and emission ranges of 520-900 and 600-1000 nm is reported. The dyes encompass commercially available rhodamine 101 (Rh-101, Φ(f) = 0.913)
Christian Würth et al.
Analytical chemistry, 84(3), 1345-1352 (2012-01-17)
There is an increasing interest in chromophores absorbing and emitting in the near-infrared (NIR) spectral region, e.g., for applications as fluorescent reporters for optical imaging techniques and hence, in reliable methods for the characterization of their signal-relevant properties like the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service