Skip to Content
Merck
All Photos(1)

Key Documents

56749

Sigma-Aldrich

Imidazole

BioUltra, ≥99.5% (GC)

Synonym(s):

1,3-Diaza-2,4-cyclopentadiene, Glyoxaline

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€80.40
250 G
€331.00
1 KG
€1,040.00

€80.40


Estimated to ship on30 March 2025


Request a Bulk Order

Select a Size

Change View
50 G
€80.40
250 G
€331.00
1 KG
€1,040.00

About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Beilstein:
103853
EC Number:
MDL number:
UNSPSC Code:
12352005
eCl@ss:
39161001
PubChem Substance ID:
NACRES:
NA.25

€80.40


Estimated to ship on30 March 2025


Request a Bulk Order

vapor pressure

<1 mmHg ( 20 °C)

Quality Level

product line

BioUltra

Assay

≥99.5% (GC)

form

powder or crystals

impurities

insoluble matter, passes filter test
≤0.2% water

ign. residue (900 °C)

≤0.05%

pH

9.5-11.0 (25 °C, 0.1 M in H2O)

pKa (25 °C)

6.95

bp

256 °C (lit.)

mp

88-91 °C (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: 0.10
λ: 280 nm Amax: 0.10

SMILES string

c1c[nH]cn1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Excellent for buffers in the range of pH 6.2-7.8
Imidazole can be used to prepare buffers in the pH range of 6.2-7.8 at 25°C. It is recommended as a component of a buffer for assay of horseradish peroxidase and as chelator for the binding of various divalent cations. Imidazole can be used for the elution of histidine containing proteins from divalent cation resins (Sigma P6611, HisSelect.-HC Nickel affinity gel) and can also be used in reverse staining of SDS-PAGE gels for detection of proteins.

Other Notes

Buffer for assay of horseradish peroxidase[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Buffer for assay of horseradish peroxidase.
B. Storrie et al.
Methods in Enzymology, 182, 217-217 (1990)
Jason Z Vlahakis et al.
Bioorganic & medicinal chemistry, 18(16), 6184-6196 (2010-07-17)
We have previously reported that tetrazolium salts were both potent and specific inhibitors of Plasmodium replication, and that they appear to interact with a parasite component that is both essential and conserved. The use of tetrazolium salts in vivo is
Aviva Levina et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(11), 3609-3619 (2013-01-31)
An anti-metastatic drug, NAMI-A ((ImH)[Ru(III) Cl4 (Im)(dmso)]; Im=imidazole, dmso=S-bound dimethylsulfoxide), and a cytotoxic drug, KP1019 ((IndH)[Ru(III) Cl4 (Ind)2 ]; Ind=indazole), are two Ru-based anticancer drugs in human clinical trials. Their reactivities under biologically relevant conditions, including aqueous buffers, protein solutions
Eleonora Petryayeva et al.
Langmuir : the ACS journal of surfaces and colloids, 29(3), 977-987 (2013-01-10)
Methods have been developed for the solid-phase detection of nucleic acids using mixed films of quantum dots (QDs) and oligonucleotide probes in microtiter plates. Polystyrene microwells were functionalized with multidentate imidazole ligands to immobilize QDs. Oligonucleotide hybridization was transduced using
Tomotsugu Awano et al.
Journal of bacteriology, 196(1), 140-147 (2013-10-29)
The genome of Thermococcus kodakarensis, along with those of most Thermococcus and Pyrococcus species, harbors five paralogous genes encoding putative α subunits of nucleoside diphosphate (NDP)-forming acyl coenzyme A (acyl-CoA) synthetases. The substrate specificities of the protein products for three

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service