Skip to Content
Merck
All Photos(1)

Key Documents

5710-OP

Millipore

Imidazole

OmniPur® Grade, >=99.0%

Synonym(s):

OmniPur® Imidazole

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
Inquire


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
500 G
Inquire

About This Item

Linear Formula:
-NHCH=NCH=CH-
CAS Number:
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.28


Estimated to ship on28 March 2025


Request a Bulk Order

Quality Level

form

crystals

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

SMILES string

[nH]1cncc1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

General description

Molecular biology grade.

Specifications:
Purity: ≥99.0%
Appearance: White to slight yellow solid
DNase: None detected
Melting point: 88-92°C
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Development of a Fluorescent Assay and Imidazole-Containing Inhibitors by Targeting SARS-CoV-2 Nsp13 Helicase.: This research develops an imidazole-containing inhibitors targeting SARS-CoV-2 Nsp13 helicase, aiding in COVID-19 therapeutic development (Zhang et al., 2024).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service