Skip to Content
Merck
All Photos(3)

Key Documents

53091-016

Sigma-Aldrich

DNAstable® Plus

2 ml, for room temp preservation of DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

grade

Molecular Biology

form

liquid

usage

80 reactions

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

SampleMatrix® by Biomatrica® is a proprietary core technology to stabilize and protect biological materials at room temperature without degradation. The SampleMatrix technology was designed by combining extremophile biology that enables long-term survival of organisms in extremely dry environments, with synthetic chemistry.

DNAstable® Plus enables you to preserve, store and ship your purified DNA samples at room temperature in liquid and liquid-to-dry formats. It offers flexibility with thermostability at extreme temperatures, and the ability to handle high concentrations of DNA. Recovery from the dry matrix is achieved within minutes by simply adding water. Your DNA sample is ready for downstream analysis without the need for any additional purification steps.

Features and Benefits

  • Long-term stabilization, storage and shipment of DNA at room temperature
  • High yield DNA recovery and use without further purification
  • Compatible with most downstream applications, e.g., PCR, sequencing, and microarrays
  • Available in a variety of compact formats, e.g., tubes, multi-well plates & custom.
  • Cost-effective, eco-friendly solution to freezer crawl and lab space management

Packaging

  • (1) DNAstable® Plus liquid reagent in a 2 mL bottle
  • Quick Reference Protocol
  • 5X solution can stabilize up to 8 mL of liquid DNA.
  • Liquid and Liquid-to-Dry (LD) stabilization
  • Liquid format is highly compatible with automation and chain-of-custody evidence preservation

Legal Information

Biomatrica is a registered trademark of Biomatrica
DNAstable is a registered trademark of Biomatrica
SampleMatrix is a registered trademark of Biomatrica

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

The product bulletin providin detailed use protocol for easy DNA transfection.

Product manual provides detailed protocol for easy DNA transfection.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service