Skip to Content
Merck
All Photos(1)

Key Documents

52926

Sigma-Aldrich

Pelargonin chloride

≥90% (HPLC)

Synonym(s):

3,5-Bis(glucosyloxy)-4′,7-dihydroxyflavylium chloride, Monardin, Pelargonidin 3,5-di-O-glucoside chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€315.00

€315.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€315.00

About This Item

Empirical Formula (Hill Notation):
C27H31ClO15
CAS Number:
Molecular Weight:
630.98
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

€315.00


Check Cart for Availability

Request a Bulk Order

biological source

synthetic

Quality Level

Assay

≥90% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[Cl-].OC[C@H]1O[C@@H](Oc2cc3c(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)cc(O)cc3[o+]c2-c5ccc(O)cc5)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C27H30O15.ClH/c28-8-17-19(32)21(34)23(36)26(41-17)39-15-6-12(31)5-14-13(15)7-16(25(38-14)10-1-3-11(30)4-2-10)40-27-24(37)22(35)20(33)18(9-29)42-27;/h1-7,17-24,26-29,32-37H,8-9H2,(H-,30,31);1H/t17-,18-,19-,20-,21+,22+,23-,24-,26-,27-;/m1./s1

InChI key

DIRROHKULXIUCB-DHJOXOLYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pelargonin is an anthocyanin, which are present in fruits and vegetables as natural colorants. It was found in dates and investigated for anti-allergic properties[1]. It shows some oxygen radical absorbance capacity[2]
The phytoestrogen pelargonin, also known as pelargonidin, is a common anthocyanin which displays antioxidant and antigenotoxic properties.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthocyanins as antioxidants on human low-density lipoprotein and lecithin-liposome systems.
Satue-Gracia, M. T., et al.
Journal of Agricultural and Food Chemistry, 45, 3362-3367 (1997)
L A Griffiths et al.
The Biochemical journal, 128(4), 901-911 (1972-07-01)
1. The metabolism of a group of flavonoid compounds related in structure to apigenin (4',5,7-trihydroxyflavone) and including apigenin, apiin, naringin, phlorrhizin, acacetin, kaempferol, robinin, chrysin, tectochrysin and 4',7-dihydroxyflavone, was studied both in vivo after oral administration to the rat, and
Koji Karasawa et al.
Journal of nutritional science and vitaminology, 58(4), 272-277 (2012-11-08)
The effects of oral ingestion of a hot water extract of matured fruit of the date palm tree (Phoenix dactylifera L.) on allergic responses were investigated in mite-sensitized mice. Sneezing and nose rubbing events in mice given a date extract-added
Oxygen radical absorbing capacity of anthocyanins.
Wang, H., et al.
Journal of Agricultural and Food Chemistry, 45, 304-309 (1997)
Gui-Fang Deng et al.
Critical reviews in food science and nutrition, 53(3), 296-306 (2012-12-12)
The pigmented rice has been consumed in China, Japan, and Korea for a long time. It has been used for strengthening kidney function, treating anemia, promoting blood circulation, removing blood stasis, treating diabetes, and ameliorating sight in traditional Chinese medicine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service