Skip to Content
Merck
All Photos(1)

Key Documents

PHL89627

Delphinidin 3-glucoside chloride

phyproof® Reference Substance

Synonym(s):

Delphinidin 3-O-β-D-glucoside chloride, Delphinidin 3-O-β-D-glucopyranoside chloride, Delphinidin 3-monoglucoside, Delphinin chloride, Myrtillin chloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€689.00

€689.00


Estimated to ship on22 March 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€689.00

About This Item

Empirical Formula (Hill Notation):
C21H21ClO12
CAS Number:
Molecular Weight:
500.84
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€689.00


Estimated to ship on22 March 2025


Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

OC1=CC(O)=C(C=C(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)C(C3=CC(O)=C(O)C(O)=C3)=[O+]4)C4=C1.[Cl-]

InChI

1S/C21H20O12.ClH/c22-6-15-17(28)18(29)19(30)21(33-15)32-14-5-9-10(24)3-8(23)4-13(9)31-20(14)7-1-11(25)16(27)12(26)2-7;/h1-5,15,17-19,21-22,28-30H,6H2,(H4-,23,24,25,26,27);1H/t15-,17-,18+,19-,21-;/m1./s1

InChI key

ZJWIIMLSNZOCBP-BTTVDUMLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

  • Delphinidin 3-glucoside chloride as a natural dye in biochemistry: Research demonstrated the potential use of Delphinidin 3-glucoside chloride as a natural dye, optimizing its application in third-generation snacks prepared using extruded blue maize, black bean, and chard, which enhances the understanding of anthocyanin stability in food products (Neder-Suárez et al., 2021).
  • Delphinidin 3-glucoside chloride antioxidant properties: Chorfa N and colleagues developed an efficient method for isolating high-purity anthocyanin isomers, including Delphinidin 3-glucoside chloride from wild blueberries, evaluating their radical scavenging activity and contributing to antioxidant biochemistry (Chorfa et al., 2016).
  • Delphinidin 3-glucoside chloride in cell signaling research: A defined anthocyanin mixture containing Delphinidin 3-glucoside chloride sourced from bilberry and black currant demonstrated inhibitory effects on Measles virus and various herpesviruses, suggesting its significant role in cellular signaling pathways related to viral inhibition (Sivarajan et al., 2022).
  • Delphinidin 3-glucoside chloride effects on cellular metabolism research: The impact of molar absorbance on the anthocyanin content of foods was studied, providing insights into how Delphinidin 3-glucoside chloride and similar compounds influence cellular metabolism, essential for polyphenol studies (Singh et al., 2022).

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service