Skip to Content
Merck
All Photos(1)

Key Documents

50935

Sigma-Aldrich

Guanidine hydrochloride

BioUltra, ≥99.5% (AT)

Synonym(s):

Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
95.53
Beilstein:
3591990
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioUltra

Assay

≥99.5% (AT)

impurities

insoluble matter, passes filter test

ign. residue

≤0.05% (as SO4)

loss

≤0.05% loss on drying, 20 °C (HV)

pH

4.5-6.0 (25 °C, 6 M in H2O)

mp

180-185 °C (lit.)

solubility

H2O: 6 M at 20 °C, clear, colorless

density

1.3 g/cm3 (lit.)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

6 M in H2O

UV absorption

λ: 260 nm Amax: 0.03
λ: 280 nm Amax: 0.02

SMILES string

Cl[H].NC(N)=N

InChI

1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H

InChI key

PJJJBBJSCAKJQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary.

Other Notes

In the extraction and fractionation by cesium sulfate density gradient centrifugation of bovine nasal cartilage proteoglycan; Induces subunit dissociation and unfolding of bovine liver glutamate dehydrogenase; Dissociation of apolipoproteins of insect lipophorin; Denaturation curve.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Tashiro et al.
Biochimica et biophysica acta, 706(1), 129-135 (1982-08-23)
Equilibrium and kinetic measurements were carried out on the denaturation of bovine liver glutamate dehydrogenase (L-glutamate:NAD(P)+ oxidoreductase (deaminating), EC 1.4.1.3) induced by guanidine hydrochloride (Gdn-HCl) in 0.2 M phosphate buffer (pH 7.3) using two types of detection, light-scattering and circular
J K Kawooya et al.
Biochemistry, 28(16), 6658-6667 (1989-08-08)
The apolipoproteins of insect lipophorin were dissociated in guanidinium chloride and isolated by gel permeation chromatography. Over 98% of the total lipid in lipophorin was associated with apolipophorin I (apoLp-I), thus suggesting this apolipoprotein to be the lipid binding component
Determination and analysis of urea and guanidine hydrochloride denaturation curves.
C N Pace
Methods in enzymology, 131, 266-280 (1986-01-01)
F Bonnet et al.
Biochimica et biophysica acta, 623(1), 57-68 (1980-05-29)
A true isopycnic centrifugation method for the study of the bovine nasal cartilage proteoglycan polydispersity is presented. The use of cesium sulfate as gradient forming salt instead of cesium chloride allowed proteoglycan banding without any sedimentation at the bottom of
Armando M De Palma et al.
Antimicrobial agents and chemotherapy, 53(5), 1850-1857 (2009-02-25)
A novel compound, TTP-8307, was identified as a potent inhibitor of the replication of several rhino- and enteroviruses. TTP-8307 inhibits viral RNA synthesis in a dose-dependent manner, without affecting polyprotein synthesis and/or processing. Drug-resistant variants of coxsackievirus B3 were all

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service