Skip to Content
Merck
All Photos(1)

Documents

37294

Sigma-Aldrich

Dihydrofolate Reductase from bovine liver

~8 U/mL, ammonium sulfate suspension, off-white

Synonym(s):

Tetrahydrofolate Dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine liver

form

ammonium sulfate suspension

specific activity

≥6 U/mg protein

concentration

~8 U/mL

color

off-white

density

1.2 g/mL at 20 °C

storage temp.

2-8°C

Application

Dihydrofolate reductase (DHFR) is involved in the synthesis of purines, thymidine and glycine in folate metabolism. DHFR, from bovine liver, is used for methotrexate analysis . DHFR is useful to study various cancers such as sarcomas, leukemias and head and neck cancers .

Biochem/physiol Actions

Dihydrofolate reductase (DHFR) reduces dihydrofolic acid to tetrahydrofolic acid, using NADPH as an electron donor. The binding of antitumor agents, such as methotrexate, to DHFR prevents the formation of reduced folates, which are essential for DNA synthesis .

Unit Definition

1 U corresponds to the amount of enzyme which converts 1 μmol 7,8-dihydrofolate and NADPH to 5,6,7,8-tetrahydrofolate and NADP per minute at pH 6.5 and 25 °C

Physical form

Suspension in 3.6 M ammonium sulfate solution, pH 7.0

Other Notes

In folate metabolism for the synthesis of purines, thymidine and glycine
Sales restrictions may apply

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H B Eastman et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(19), 8572-8576 (1991-10-01)
Dihydrofolate reductase (DHFR; EC 1.5.1.3) is required in folate metabolism for the synthesis of purines, thymidine, and glycine. Although there have been several reports of induction of DHFR enzyme by methotrexate (MTX), a drug that competitively inhibits DHFR, there are
Use of dihydrofolate reductase from bovine liver, rather than from L. casei, for determining methotrexate.
R J Brooks
Clinical chemistry, 24(3), 518-519 (1978-03-01)
Tetsuo Shimizu et al.
Anticancer research, 32(10), 4589-4596 (2012-10-13)
Pemetrexed inhibits three key folate enzymes: thymidylate synthetase (TYMS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT). The relationship between the clinical efficacy of pemetrexed and the expression of folate enzymes in lung cancer cells is unknown. The purpose of
Luca Belmonte et al.
Critical reviews in eukaryotic gene expression, 22(3), 219-232 (2012-11-13)
A state-of-the-art review of the role of the Langmuir-Blodgett nanotemplate on protein crystal structures is here presented. Crystals grown by nanostructured template appear more radiation resistant than the classical ones, even in the presence of a third-generation highly focused beam
S P Chumakov et al.
Molekuliarnaia biologiia, 46(5), 699-711 (2012-11-20)
Association and degradation of protein complexes play essential role in a majority of normal and pathologic processes, which take place in living cell. Studying the underlying mechanisms of those interactions would give deeper understanding of specific causes of disease progression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service