Merck
  • Association between expression of thymidylate synthase, dihydrofolate reductase, and glycinamide ribonucleotide formyltransferase and efficacy of pemetrexed in advanced non-small cell lung cancer.
Association between expression of thymidylate synthase, dihydrofolate reductase, and glycinamide ribonucleotide formyltransferase and efficacy of pemetrexed in advanced non-small cell lung cancer.
Anticancer research (2012-10-13)
Tetsuo Shimizu, Yoko Nakanishi, Yoshiko Nakagawa, Ichiro Tsujino, Noriaki Takahashi, Norimichi Nemoto, Shu Hashimoto
ABSTRACT
Pemetrexed inhibits three key folate enzymes: thymidylate synthetase (TYMS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT). The relationship between the clinical efficacy of pemetrexed and the expression of folate enzymes in lung cancer cells is unknown. The purpose of this study was to determine whether TYMS, DHFR, and GARFT expression affect the therapeutic efficacy of pemetrexed. Participants (n=50) were patients with advanced non-small cell lung cancer (NSCLC) treated with pemetrexed. Samples were obtained by tumor biopsy before treatment. We isolated cancer cells from formalin-fixed paraffin-embedded tissues using laser microdissection, and mRNA levels were analyzed using real-time reverse transcription polymerase chain reaction. Protein expression was evaluated using immunohistochemistry. We assessed the association between TYMS, DHFR, and GARFT expression and the therapeutic efficacy of pemetrexed. The median age was 66.8 years. Compared to healthy tissues, the relative TYMS mRNA expression ranged from 0.001 to 41.613 (mean 4.638 ± 1.357), and was significantly lower in responders compared to non-responders (1.671 ± 0.844 versus 5.978 ± 1.895, p=0.0142). Progression-free survival was prolonged in patients with lower TYMS mRNA expression compared to those with higher TYMS mRNA expression, but the difference was not statistically significant (18.0 versus 13.3 weeks, p=0.3001). DHFR and GARFT mRNA expression did not correlate with the efficacy of pemetrexed. We specifically analyzed TYMS, DHFR, and GARFT mRNA expression levels in lung cancer cells from biopsy specimens using laser microdissection. TYMS mRNA expression affected the therapeutic efficacy of pemetrexed and could therefore constitute a useful predictive biomarker for NSCLC patients receiving pemetrexed.
Sigma-Aldrich
Dihydrofolate Reductase human, ≥80% (SDS-PAGE), recombinant, expressed in E. coli, ≥1 units/mg protein
Sigma-Aldrich
Dihydrofolate Reductase from bovine liver, ~8 U/mL, ammonium sulfate suspension, off-white