Skip to Content
Merck
All Photos(1)

Key Documents

36900

Supelco

PCB No 28 solution

10 μg/mL in isooctane, analytical standard

Synonym(s):

2,4,4′-Trichlorobiphenyl solution, 2,4,4′-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7Cl3
CAS Number:
Molecular Weight:
257.54
Beilstein:
1956846
Ballschmiter Number:
28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in isooctane

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)-c2ccc(Cl)cc2Cl

InChI

1S/C12H7Cl3/c13-9-3-1-8(2-4-9)11-6-5-10(14)7-12(11)15/h1-7H

InChI key

BZTYNSQSZHARAZ-UHFFFAOYSA-N

General description

Polychlorinated biphenyls (PCBs) belong to the class of highly toxic pollutants commonly found in various environmental matrices and food products. They find a variety of applications as flame retardants, plasticizers, dielectric and heat transfer fluids, etc.[1]

Application

PCB No 28 solution may be used as an analytical reference standard for the determination of the analyte in water samples using gas chromatography coupled to mass spectrometry (GC-MS).[2]

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polychlorinated Biphenyl Compounds (PCBs) Determination from Water by Gas Chromatography Coupled with Mass Spectrometry (GC?MS)
Senin R, et al.
Revista de Chimie (Bucharest, Romania), 67(11), 2134-2140 (2016)
Xiang-Hui Sun et al.
Huan jing ke xue= Huanjing kexue, 31(10), 2327-2330 (2011-01-15)
Sorption behaviors of PCB28 to phosphatidylcholine (PC)and triglyceride (TG) were studied. Results showed that sorption equilibrium could be achieved in 8 h for PCB28, and the sorption amount on PC was higher than that of TG when the initial PCB28
Li Xu et al.
Huan jing ke xue= Huanjing kexue, 31(1), 255-259 (2010-03-25)
The capability of Rhizobium meliloti on degrading 2,4,4'-trichlorobiphenyl and 18 kinds of polychlorinated biphenyl congener mixtures was studied by shaking flask experiment. The results showed that the degradation capability of Rhizobium meliloti to 2, 4, 4'-trichlorobiphenyl increased gradually with the
Isolation and characterisation of polychlorinated biphenyl (PCB) degrading fungi from a historically contaminated soil
Tigini V, et al.
Microbial cell factories, 8(1), 5-5 (2009)
Chen Tu et al.
Journal of hazardous materials, 186(2-3), 1438-1444 (2011-01-05)
Resting cell assay and soil microcosms were set up to investigate the biodegradation capability and metabolic intermediate of polychlorinated biphenyls (PCBs) by a rhizobial strain Sinorhizobium meliloti. Biodegradation was observed immediately after 2,4,4'-TCB was supplied as a sole source of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service