Skip to Content
Merck
All Photos(1)

Key Documents

2851

Sigma-Aldrich

Citrate Phosphate Buffer Concentrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€105.00

€105.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€105.00

About This Item

UNSPSC Code:
41116124
NACRES:
NA.47

€105.00


Estimated to ship on28 May 2025


Request a Bulk Order

form

solution

IVD

for in vitro diagnostic use

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Application


  • Poly(sulfobetaine) versus poly(ethylene glycol) based copolymer modified polyurethane catheters for antifouling: This study examines the use of ethanol in the synthesis of polymers designed to reduce biological fouling on medical devices, highlighting its role in advanced healthcare materials research (Cao et al., 2024).

  • Green synthesis of polyimide by using an ethanol solvothermal method for aqueous zinc batteries: Ethanol is used as a green solvent in the innovative synthesis of polyimide structures, emphasizing its importance in sustainable material production and energy storage solutions (Zhao et al., 2024).

  • Modulation of gut microbiota with probiotics as a strategy to counteract endogenous and exogenous neurotoxicity: Research explores the indirect applications of ethanol in the solvent extraction processes used for studying neuroprotective strategies in medical research (Skalny et al., 2024).

  • Superselective Ablative Chemo-ethanol Embolization for Recurrent Single Hepatocellular Carcinoma: A Six-Month Outcome Analysis: This clinical study utilizes ethanol directly in therapeutic interventions, demonstrating its critical use in oncological treatments to improve patient outcomes (Lee et al., 2024).

  • Self-Assembly Reinforced Alginate Fibers for Enhanced Strength, Toughness, and Bone Regeneration: Ethanol is employed in the processing of novel biomaterials designed for medical applications, showcasing its utility in the development of regenerative medicine technologies (Cui et al., 2024).

Preparation Note

Sodium citrate, 0.7 mol/L, and sodium phosphate, 0.6 mol/L

Disclaimer

Sales restrictions may apply

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Are adult red blood cells failing to properly elute hemoglobin after incubation in the Citrate Phosphate Buffer working solution? Could the pH of the Citrate Phosphate Buffer be too high? Additionally, is it feasible to increase the amount of agitation to aid in eluting the adult hemoglobin?

    1 answer
    1. To elute adult hemoglobin, pure/undenatured 80% alcohol is necessary. The use of histological or reagent grade alcohol may result in inadequate elution.

      Helpful?

  2. Is it advisable to switch to the published procedure, which requires a 5-minute incubation, considering the change in cell appearance after immersing the slides for 2 minutes in the buffer?

    1 answer
    1. The recommended procedure for a 5-minute incubation in 1X Citrate Phosphate Buffer solution should be followed, with adjustments made based on the degree of staining observed with the slides. The slides should be agitated at 1 and 3 minutes, and the user should determine the amount of agitation required based on their observations. The degree of agitation may need to be adjusted to address issues such as hard-to-observe ghost cells or an excess of adult hemoglobin.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service