Skip to Content
Merck
All Photos(1)

Key Documents

27042

Sigma-Aldrich

Chondroitin 4-sulfate sodium salt from bovine trachea

mixture of isomers, main component: chondroitin 4-sulfate

Synonym(s):

CSA, Chondroitin sulfate A sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

bovine trachea

form

powder

optical activity

[α]20/D −25±5°, c = 1% in H2O

impurities

≤10% water

solubility

H2O: 0.1 g/mL, clear to slightly turbid, colorless to light yellow

storage temp.

2-8°C

SMILES string

CC1=C(CCN2CCC(\CC2)=C(/c3ccccc3)c4ccc(F)cc4)C(=O)N5C=CSC5=N1

Application

Chondroitin, a glucosaminoglycan (GAG), is a polysaccharide chain of alternating units of N-acetylgalactosamine (GalNAc) and glucuronic acid (GlcA) that can be sulfated on the either or both GalNAc and GlcA units. Chondroitin and its sulfates are frequently attached to proteins to form proteoglycans.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clare M Galtrey et al.
Brain research reviews, 54(1), 1-18 (2007-01-16)
Chondroitin sulfate proteoglycans (CSPGs) consist of a core protein and glycosaminoglycan (GAG) chains. There is enormous structural diversity among CSPGs due to variation in the core protein, the number of GAG chains and the extent and position of sulfation. Most
Tomomi Izumikawa et al.
Biochimica et biophysica acta, 1830(10), 4806-4812 (2013-07-03)
Previously, we identified two missense mutations in the chondroitin N-acetylgalactosaminyltransferase-1 gene in patients with neuropathy. These mutations are associated with a profound decrease in chondroitin N-acetylgalactosaminyltransferase-1 enzyme activity. Here, we describe a patient with neuropathy who is heterozygous for a
Gunnar Dick et al.
The Journal of biological chemistry, 288(38), 27384-27395 (2013-08-14)
Chondroitin sulfate (CS) and the CS-rich extracellular matrix structures called perineuronal nets (PNNs) restrict plasticity and regeneration in the CNS. Plasticity is enhanced by chondroitinase ABC treatment that removes CS from its core protein in the chondroitin sulfate proteoglycans or
J C F Kwok et al.
The international journal of biochemistry & cell biology, 44(4), 582-586 (2012-01-24)
Chondroitin sulfate is a glycosaminoglycan composed of N-acetylgalactosamine and glucuronic acid. It attaches to a core protein to form chondroitin sulfate proteoglycan (CSPG). Being a major component of the brain extracellular matrix, CSPGs are involved in neural development, axon pathfinding
Stella Bauer et al.
Langmuir : the ACS journal of surfaces and colloids, 29(12), 4039-4047 (2013-02-22)
Polysaccharides are a promising material for nonfouling surfaces because their chemical composition makes them highly hydrophilic and able to form water-storing hydrogels. Here we investigated the nonfouling properties of hyaluronic acid (HA) and chondroitin sulfate (CS) against marine fouling organisms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service