Skip to Content
Merck
All Photos(1)

Key Documents

230699

Millipore

Chondroitin sulfate from bovine trachea

Fundamental component of connective tissue. Acts as a flexible connecting matrix between tough protein filaments in cartilage to form a polymeric system. Mixture of chondroitin sulfates A, C, D, and E.

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€136.00

€136.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€136.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79

€136.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 2214

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O[C@H]1O[C@H]([C@@H]([C@H]([C@H]1O)O[C@@H]2O[C@@H]([C@H]([C@@H]([C@H]2O)O)O)C(=O)O)NC(=O)C)O)O

InChI

1S/C13H21NO15S/c1-2(15)14-3-8(7(19)13(28-11(3)22)29-30(23,24)25)26-12-6(18)4(16)5(17)9(27-12)10(20)21/h3-9,11-13,16-19,22H,1H3,(H,14,15)(H,20,21)(H,23,24,25)/t3-,4+,5+,6-,7-,8-,9+,11-,12-,13-/m1/s1

InChI key

KXKPYJOVDUMHGS-OSRGNVMNSA-N

General description

Fundamental component of connective tissue. Acts as a flexible connecting matrix between tough protein filaments in cartilage to form a polymeric system. Mixture of chondroitin sulfates A, C, D, and E.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service