Skip to Content
Merck
All Photos(2)

Key Documents

16328

Sigma-Aldrich

O-[2-(Boc-amino)ethyl]-O′-(2-maleimidoethyl)ethylene glycol

≥96.0% (HPLC)

Synonym(s):

tert-Butyl {2-{2-[2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethoxy]ethoxy}ethyl}carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€807.00

€807.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€807.00

About This Item

Empirical Formula (Hill Notation):
C15H24N2O6
CAS Number:
Molecular Weight:
328.36
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

€807.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥96.0% (HPLC)

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCOCCOCCN1C(=O)C=CC1=O

InChI

1S/C15H24N2O6/c1-15(2,3)23-14(20)16-6-8-21-10-11-22-9-7-17-12(18)4-5-13(17)19/h4-5H,6-11H2,1-3H3,(H,16,20)

InChI key

ANPNLGYUZZQPEV-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service