Skip to Content
Merck
All Photos(2)

Documents

226351

Sigma-Aldrich

N-Hydroxymaleimide

97%, for peptide synthesis

Synonym(s):

1-Hydroxypyrrole-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3NO3
CAS Number:
Molecular Weight:
113.07
Beilstein:
1524594
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

product name

N-Hydroxymaleimide, 97%

Assay

97%

form

solid

reaction suitability

reaction type: Addition Reactions

mp

130-133 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

ON1C(=O)C=CC1=O

InChI

1S/C4H3NO3/c6-3-1-2-4(7)5(3)8/h1-2,8H

InChI key

BUXKULRFRATXSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Hydroxymaleimide (NHMI) is an organic compound that is widely used as an oxidizing agent in organic synthesis. It is frequently used to selectively oxidize thiols to disulfides and in the synthesis of α, β-unsaturated carbonyl compounds. It can also act as a Michael acceptor and is used in the synthesis of peptides and proteins.

Application

N-Hydroxymaleimide can be used as an oxidant to generate N-hydroxy anilines in situ, which serve as key intermediates in the selective coupling reaction of N-substituted anilines with hydroxylamine derivatives, for the efficient synthesis of aromatic oximes.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minye Yang et al.
Nanoscale, 10(33), 15865-15874 (2018-08-15)
The detection of mycotoxins in food is urgently needed because they pose a significant threat to public health. In this study, we developed a quantitative detection platform for mycotoxins by integrating multicolor upconversion nanoparticle barcode technology with fluorescence image processing
Jamerson Carneiro de Oliveira et al.
Molecules (Basel, Switzerland), 25(2) (2020-01-16)
The study of Diels-Alder reactions in materials science is of increasing interest. The main reason for that is the potential thermoreversibility of the reaction. Aiming to predict the behavior of a material modified with maleimido and furyl moieties, 1H NMR
Yusuke Hibi et al.
Nature communications, 7, 11064-11064 (2016-03-22)
There is a growing interest in sequence-controlled polymers toward advanced functional materials. However, control of side-chain order for vinyl polymers has been lacking feasibility in the field of polymer synthesis because of the inherent feature of chain-growth propagation. Here we
Moon Soo Gil et al.
Journal of controlled release : official journal of the Controlled Release Society, 267, 119-132 (2017-04-17)
Biological drugs are exquisitely tailored components offering the advantages of high specificity and efficacy that are considered safe for treating diseases. Nevertheless, the effectiveness of biological drugs is limited by their inherent short biological half-life and poor stability in vivo.
Jinhwan Cho et al.
Journal of controlled release : official journal of the Controlled Release Society, 324, 532-544 (2020-05-27)
Therapeutic proteins are attractive candidates for the treatment of human diseases. However, their short half-life often limits their clinical application. To overcome this problem, injectable hydrogels have been developed as depots for controlled release of therapeutic proteins, but these systems

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service