Skip to Content
Merck
All Photos(1)

Key Documents

121-25-02

Sigma-Aldrich

Extracellular Matrix Protein Coated T-25 Flasks

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€50.00

€50.00


Estimated to ship on28 May 2025



Select a Size

Change View
1 EACH
€50.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

€50.00


Estimated to ship on28 May 2025


manufacturer/tradename

Cell Applications, Inc

Quality Level

surface area

25 cm2

matrix active group

ECM Protein coating

shipped in

wet ice

storage temp.

2-8°C

General description

T-25 tissue culture flasks pre-coated with Extracellular Matrix Attachment Factor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

Technical information for working with human lung microvascular endothelial cells including thawing, subculturing and cryopreservation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service