Skip to Content
Merck
All Photos(3)

Key Documents

34858

Sigma-Aldrich

Ethyl acetate

suitable for HPLC, ≥99.7%

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.7%
technique(s):
GC/MS: suitable
HPLC: suitable
application(s):
food and beverages
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)

Agency

suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b

Quality Level

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

Assay

≥99.7%

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

technique(s)

GC/MS: suitable
HPLC: suitable

impurities

≤0.0005% non-volatile matter
≤0.005% free acid (as CH3COOH)
≤0.03% water (Karl Fischer)

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

UV absorption

λ: 255 nm Amax: ≤0.70
λ: 260 nm Amax: ≤0.30
λ: 270 nm Amax: ≤0.10
λ: 300 nm Amax: ≤0.01
λ: 400 nm Amax: ≤0.01

application(s)

food and beverages

format

neat

storage temp.

room temp

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl acetate is an effective alternate solvent of diethyl ether.

Application

Ethyl acetate has been used as solvent for the isolation of Rose hip (Rosa canina L., Rosaceae) powder, via sonication. It has been used as solvent for the abstraction of volatile thiols from wine for their quantitative estimation by gas chromatography/mass spectrometry (GC-MS).

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.
Important notice
  • The article number 34858-4X2.5L will be discontinued. Please order the single bottle 34858-2.5L which is physically identical with the same exact specifications.
  • The article number 34858-6X1L will be discontinued. Please order the single bottle 34858-1L which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Maria Nikolantonaki et al.
Analytica chimica acta, 660(1-2), 102-109 (2010-01-28)
As volatile thiols are nucleophiles, they are capable of additional reactions with electrophiles. In enology, this concerns reactions between volatile or non-volatile thiols and oxidized phenolic compounds. Initial studies concerning the reactivity of volatile thiols with polyphenols showed that (+)-catechin
Lasse Saaby et al.
Phytotherapy research : PTR, 25(2), 195-201 (2010-07-16)
A previously published systematic review and a metaanalysis have concluded that the consumption of standardized rose hip powder (Rosa canina L.) can reduce pain in osteoarthritis patients. Synovial inflammation has been suggested to play an important role in the pathogenesis
K H Young et al.
Journal of clinical microbiology, 10(6), 852-853 (1979-12-01)
Ethyl acetate appears to be a satisfactory subsitute solvent for diethyl ether in the Formalin-ether sedimentation technique. In comparative studies, concentration of organisms with ethyl acetate was equal to or greater than that with diethyl ether. No distortion or alteration
Flavio G Rocha et al.
Biomaterials, 29(19), 2884-2890 (2008-04-09)
Our group has previously created a functional neointestine that is capable of restoring absorptive function. However, the endogenous level of vascular endothelial growth factor (VEGF) is markedly reduced in the construct compared to native bowel. Therefore, we wanted to locally
Tsukasa Horiyama et al.
PloS one, 9(9), e108642-e108642 (2014-09-27)
Escherichia coli produces the iron-chelating compound enterobactin to enable growth under iron-limiting conditions. After biosynthesis, enterobactin is released from the cell. However, the enterobactin export system is not fully understood. Previous studies have suggested that the outer membrane channel TolC

Articles

Rapid & accurate analysis of 208 pesticides and their metabolites in a fruit sample using Supel™ QuE QuEChERS mixes for sample preparation and GC-MS/MS.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service