Skip to Content
Merck
All Photos(1)

Key Documents

1.07048

Supelco

Ethyl acetate

Synonym(s):

Ethyl ethanoate, Acetic ester, Ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
€162.00

€162.00


Estimated to ship on02 April 2025


Request a Bulk Order

Select a Size

Change View
4 L
€162.00

About This Item

UNSPSC Code:
12352108
NACRES:
NA.05
grade:
ACS reagent
for analytical purposes
for extraction
reagent grade
Assay:
≥99.5% (GC)
application(s):
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation
bp:
77 °C/1013 hPa
vapor pressure:
97 hPa ( 20 °C)

€162.00


Estimated to ship on02 April 2025


Request a Bulk Order

grade

ACS reagent
for analytical purposes
for extraction
reagent grade

Quality Level

vapor pressure

97 hPa ( 20 °C)

product line

EMPARTA®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

460 °C

potency

5620 mg/kg LD50, oral (rat)
>18000 mg/kg LD50, skin (rabbit)

expl. lim.

2.1-11.5 % (v/v)

bp

77 °C/1013 hPa

mp

-83 °C

transition temp

flash point -4 °C

solubility

85.3 g/L

density

0.9 g/cm3 at 20 °C

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

storage temp.

2-25°C

General description

for analysis Emparta® ACS

Application


  • Anti-senescence effects of Rhodiola crenulate extracts on LO(2) cells and bioactive compounds.: This study highlights the use of ethyl acetate in the extraction of compounds from Rhodiola crenulate, demonstrating significant anti-aging effects on liver cells, which can be pivotal in analytical chemistry for age-related research and drug development (Liang et al., 2023).

  • Saccorhiza polyschides-A Source of Natural Active Ingredients for Greener Skincare Formulations.: Discusses the extraction of natural ingredients using ethyl acetate, contributing to the development of greener and more sustainable skincare products, underlining the extraction and analysis methods in green chemistry (Susano et al., 2022).

Storage and Stability

Store at +2°C to +25°C.

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service