Skip to Content
Merck
All Photos(1)

Key Documents

31483

Sigma-Aldrich

Nickel(II) sulfate hexahydrate

puriss. p.a., ACS reagent, 99-102.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiSO4 · 6H2O
CAS Number:
Molecular Weight:
262.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form:
crystals
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

vapor density

2.07 (vs air)

Assay

99-102.0%

form

crystals

impurities

≤0.001% total nitrogen (N)
≤0.005% in water insoluble matters

pH

4-6 (20 °C, 5%)

anion traces

chloride (Cl-): ≤10 mg/kg

cation traces

Ca: ≤50 mg/kg
Co: ≤20 mg/kg
Cu: ≤20 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤5 mg/kg
Na: ≤100 mg/kg
Pb: ≤10 mg/kg
Zn: ≤20 mg/kg

suitability

complies for identity (IR)

SMILES string

O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

InChI

1S/Ni.H2O4S.6H2O/c;1-5(2,3)4;;;;;;/h;(H2,1,2,3,4);6*1H2/q+2;;;;;;;/p-2

InChI key

RRIWRJBSCGCBID-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) sulfate hexahydrate is a hydrated nickel salt. Its thermal decomposition in the presence of air and helium has been studied.[1] Its crystals exist in monoclinic and tetragonal polymorphic forms.[2]

Application

Nickel(II) sulfate hexahydrate may be used in the preparation of NiFe2O4 nanoparticles (NPs), which are required for the fabrication of a glucose biosensor.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amperometric glucose biosensor based on NiFe2O4 nanoparticles and chitosan.
Luo L, et al.
Sensors and Actuators B, Chemical, 145(!), 293-298 (2010)
Lan Chen et al.
International journal of molecular sciences, 22(5) (2021-03-07)
Nickel (Ni2+) is one of the most common allergens, affecting around 10-15% of the general population. As the demand for orthopedic implant surgery rises, the number of surgical revisions due to joint implant failure also increases. There is evidence that
Polymorphism of nickel sulfate hexahydrate.
Angel RJ and Finger LW.
Acta Crystallographica Section C, Crystal Structure Communications, 44(11), 1869-1873 (1988)
Mechanism and kinetics of thermal decomposition of nickel (II) sulfate (VI) hexahydrate.
Tomaszewicz E and Kotfica M.
Journal of Thermal Analysis and Calorimetry, 77(1), 25-31 (2004)
A Tammaro et al.
International journal of immunopathology and pharmacology, 25(2), 519-521 (2012-06-16)
Nickel deficiency leads to reduced iron content in organs and to reduced hemoglobin levels and haematocrit counts. We carried out a clinical trial of oral hyposensitization therapy with low doses of nickel on a group of 20 patients affected by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service