Skip to Content
Merck
All Photos(1)

Key Documents

1.06726

Supelco

Nickel(II) sulfate hexahydrate

EMPLURA®

Synonym(s):

Nickel(II) sulfate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
262.86
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-104-9
NACRES:
NA.21
form:
crystals
solubility:
650 g/L
Pricing and availability is not currently available.

Quality Level

product line

EMPLURA®

Assay

≥98.0% (complexometric)

form

crystals

potency

361 mg/kg LD50, oral (rat)

impurities

≤0.5% Substances not precipitated by ammonium sulfide (as SO4)

pH

4.3-4.7 (20 °C, 100 g/L in H2O)

solubility

650 g/L

density

2.07 g/cm3 at 20 °C

bulk density

1000 kg/m3

anion traces

chloride (Cl-): ≤0.003%

cation traces

Co: ≤0.05%
Fe: ≤0.01%
Pb: ≤0.001%
Zn: ≤0.2%

storage temp.

2-30°C

SMILES string

[Ni+2].[S](=O)(=O)([O-])[O-].O.O.O.O.O.O

InChI

1S/Ni.H2O4S.6H2O/c;1-5(2,3)4;;;;;;/h;(H2,1,2,3,4);6*1H2/q+2;;;;;;;/p-2

InChI key

RRIWRJBSCGCBID-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

EMPLURA®

Analysis Note

Assay (complexometric)≥ 98.0 %Identitypasses testChloride (Cl)≤ 0.003 %Co (Cobalt)≤ 0.05 %Fe (Iron)≤ 0.01 %Pb (Lead)≤ 0.001 %Zn (Zinc)≤ 0.2 %Substances not precipitated by ammonium sulfide (as SO₄)≤ 0.5 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service